Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp2cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213293.1 | NP_998458.1 | 6 - 7 | 1166 | 126 | |
GenBank mRNA | BC044495.1 | - | 6 - 7 | 1131 | 126 | |
BC065680.1 | - | 6 - 7 | 1166 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |