Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pias4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213403.2 | NP_998568.2 | 6 - 7 | 777 | 104 | |
GenBank mRNA | BC057528.1 | - | 6 - 7 | 767 | 104 | |
BC070017.1 | - | 6 - 7 | 777 | 104 | ||
JF759916.1 | - | 6 - 7 | 796 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |