Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pias4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213403.2 | NP_998568.2 | 7 - 8 | 872 | 111 | |
GenBank mRNA | BC057528.1 | - | 7 - 8 | 862 | 111 | |
BC070017.1 | - | 7 - 8 | 872 | 111 | ||
JF759916.1 | - | 7 - 8 | 891 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |