Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tph2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214795.2 | NP_999960.2 | 1 - 2 | 330 | 90 | |
GenBank mRNA | AB125219.1 | - | 1 - 2 | 330 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |