Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: polr2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002317.2 | NP_001002317.2 | 2 - 3 | 365 | 91 | |
GenBank mRNA | AY648795.1 | - | 2 - 3 | 345 | 91 | |
BC095261.1 | - | 2 - 3 | 259 | 91 | ||
BC164278.1 | - | 2 - 3 | 290 | 91 | ||
EH541343.1 | - | 2 - 3 | 405 | 91 | ||
EH555518.1 | - | 2 - 3 | 402 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |