Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc25a48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002367.1 | NP_001002367.1 | 2 - 3 | 199 | 147 | |
XM_009291023.3 | XP_009289298.1 | 2 - 3 | 262 | 170 | ||
XM_021481000.1 | XP_021336675.1 | 2 - 3 | 267 | 147 | ||
GenBank mRNA | BC075888.1 | - | 2 - 3 | 199 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |