Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc25a48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002367.1 | NP_001002367.1 | 3 - 4 | 275 | 105 | |
XM_021481000.1 | XP_021336675.1 | 3 - 4 | 343 | 105 | ||
GenBank mRNA | BC075888.1 | - | 3 - 4 | 275 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |