Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fhl2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003732.1 | NP_001003732.1 | 4 - 5 | 545 | 68 | |
XM_005167436.4 | XP_005167493.1 | 5 - 6 | 1080 | 68 | ||
GenBank mRNA | BC078393.1 | - | 4 - 5 | 545 | 68 | |
DQ118096.1 | - | 3 - 4 | 515 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |