Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gspt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003992.1 | NP_001003992.1 | 5 - 6 | 725 | 96 | |
GenBank mRNA | BC080263.1 | - | 5 - 6 | 725 | 96 | |
BC164587.1 | - | 5 - 6 | 591 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |