Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tfr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009917.1 | NP_001009917.1 | 6 - 7 | 889 | 107 | |
XM_005163666.4 | XP_005163723.1 | 7 - 8 | 990 | 107 | ||
GenBank mRNA | AY649363.1 | - | 6 - 7 | 889 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |