Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bnip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012245.1 | NP_001012245.1 | 4 - 5 | 476 | 89 | |
XM_005172968.4 | XP_005173025.1 | 4 - 5 | 749 | 89 | ||
GenBank mRNA | AY424950.1 | - | 4 - 5 | 476 | 89 | |
BC171679.1 | - | 4 - 5 | 440 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |