Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: drd4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012618.1 | NP_001012636.1 | 3 - 4 | 961 | 70 | |
GenBank mRNA | AY750154.1 | - | 3 - 4 | 961 | 70 | |
BC162965.1 | - | 3 - 4 | 1041 | 70 | ||
BC162989.1 | - | 3 - 4 | 1041 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |