Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: asrgl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013529.1 | NP_001013547.1 | 4 - 5 | 582 | 76 | |
GenBank mRNA | BC090901.1 | - | 4 - 5 | 582 | 76 | |
BC164091.1 | - | 3 - 4 | 515 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |