Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tvp23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024428.1 | NP_001019599.1 | 4 - 5 | 369 | 87 | |
GenBank mRNA | BC095132.1 | - | 4 - 5 | 369 | 87 | |
BC165009.1 | - | 3 - 4 | 357 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |