Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: apol1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030138.1 | NP_001025309.1 | 1 - 2 | 61 | 83 | |
XM_005171301.3 | XP_005171358.1 | 8 - 9 | 705 | 83 | ||
XM_005171302.3 | XP_005171359.1 | 8 - 9 | 702 | 83 | ||
XM_005171303.3 | XP_005171360.1 | 7 - 8 | 630 | 83 | ||
GenBank mRNA | BC115193.1 | - | 1 - 2 | 51 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |