Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scara5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030190.1 | NP_001025361.1 | 5 - 6 | 1001 | 95 | |
XM_005160318.4 | XP_005160375.1 | 6 - 7 | 1303 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |