Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: olah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039932.1 | NP_001035021.1 | 3 - 4 | 574 | 81 | |
XM_005159455.3 | XP_005159512.1 | 3 - 4 | 356 | 81 | ||
XM_021468123.1 | XP_021323798.1 | 3 - 4 | 578 | 81 | ||
GenBank mRNA | BC114310.1 | - | 3 - 4 | 574 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |