Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chrna10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044804.1 | NP_001038269.1 | 2 - 3 | 353 | 92 | |
XM_021481417.1 | XP_021337092.1 | 3 - 4 | 501 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Danio rerio Probe spans exons |