Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pmaip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045474.3 | NP_001038939.1 | 2 | 157 | 100 | |
GenBank mRNA | BC134980.1 | - | 2 | 157 | 100 | |
DQ860152.1 | - | 1 | 69 | 100 | ||
EH549974.1 | - | 2 | 199 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |