Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lect2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048055.1 | NP_001041520.1 | 2 - 3 | 188 | 148 | |
GenBank mRNA | BC162786.1 | - | 2 - 3 | 125 | 148 | |
BC162799.1 | - | 2 - 3 | 125 | 148 | ||
DQ372604.1 | - | 2 - 3 | 188 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |