Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdpk1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077344.1 | NP_001070812.1 | 5 - 6 | 765 | 98 | |
GenBank mRNA | BC091466.1 | - | 6 - 7 | 707 | 98 | |
BC124736.1 | - | 5 - 6 | 765 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |