Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: drc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077727.1 | NP_001071195.1 | 9 - 10 | 1172 | 105 | |
XM_021471579.1 | XP_021327254.1 | 9 - 10 | 1181 | 105 | ||
GenBank mRNA | BC125885.1 | - | 9 - 10 | 1172 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |