Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plekhm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001327851.1 | NP_001314780.1 | 5 - 6 | 1560 | 96 | |
NM_001327852.1 | NP_001314781.1 | 4 - 5 | 921 | 96 | ||
XM_021480128.1 | XP_021335803.1 | 5 - 6 | 1707 | 96 | ||
GenBank mRNA | BC133733.1 | - | 4 - 5 | 872 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |