Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: exoc3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099422.1 | NP_001092892.1 | 4 - 5 | 932 | 69 | |
XM_005174338.4 | XP_005174395.1 | 4 - 5 | 1692 | 69 | ||
GenBank mRNA | BC139871.1 | - | 4 - 5 | 932 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |