Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cmbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109832.1 | NP_001103302.1 | 3 - 4 | 407 | 108 | |
XM_009297347.3 | XP_009295622.1 | 3 - 4 | 588 | 108 | ||
GenBank mRNA | BC151924.1 | - | 3 - 4 | 407 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |