Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cmbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109832.1 | NP_001103302.1 | 4 - 5 | 554 | 92 | |
XM_009297347.3 | XP_009295622.1 | 4 - 5 | 735 | 92 | ||
GenBank mRNA | BC151924.1 | - | 4 - 5 | 554 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |