Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btbd10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001347897.1 | NP_001334826.1 | 2 - 3 | 307 | 106 | |
NR_144779.1 | 2 - 3 | 307 | 106 | |||
GenBank mRNA | BC134926.1 | - | 2 - 3 | 270 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |