Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btbd10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_144779.1 | 9 - 10 | 1779 | 67 | ||
GenBank mRNA | BC134926.1 | - | 9 - 10 | 1730 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |