Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131152.1 | NP_571227.1 | 3 - 4 | 264 | 88 | |
GenBank mRNA | AF032392.1 | - | 3 - 4 | 264 | 88 | |
BC092699.1 | - | 3 - 4 | 310 | 88 | ||
BC165062.1 | - | 3 - 4 | 325 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |