Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: etv5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309209.1 | NP_001296138.1 | 6 - 7 | 564 | 92 | |
NM_131205.2 | NP_571280.2 | 6 - 7 | 564 | 92 | ||
XM_005165860.4 | XP_005165917.1 | 6 - 7 | 584 | 92 | ||
XM_005165861.4 | XP_005165918.1 | 6 - 7 | 584 | 92 | ||
GenBank mRNA | AF168007.1 | - | 5 - 6 | 447 | 92 | |
BC060924.1 | - | 6 - 7 | 545 | 92 | ||
CT594467.2 | - | 2 - 3 | 140 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |