Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: foxh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131502.1 | NP_571577.1 | 1 - 2 | 619 | 90 | |
XM_005156080.4 | XP_005156137.1 | 2 - 3 | 815 | 90 | ||
GenBank mRNA | AF263000.1 | - | 1 - 2 | 619 | 90 | |
AF264751.1 | - | 1 - 2 | 367 | 90 | ||
BC044340.1 | - | 1 - 2 | 630 | 90 | ||
BC164213.1 | - | 1 - 2 | 401 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |