Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tram1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153669.2 | NP_705955.2 | 9 - 10 | 957 | 115 | |
GenBank mRNA | AY029529.1 | - | 9 - 10 | 966 | 115 | |
BC052143.1 | - | 9 - 10 | 1017 | 115 | ||
BC063238.1 | - | 9 - 10 | 957 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |