Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nifk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173288.1 | NP_775395.1 | 1 - 2 | 222 | 71 | |
GenBank mRNA | AF506200.1 | - | 1 - 2 | 222 | 71 | |
BC071545.1 | - | 1 - 2 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |