Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smad3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175083.2 | NP_778258.1 | 7 - 8 | 1016 | 69 | |
GenBank mRNA | AY134490.1 | - | 7 - 8 | 1016 | 69 | |
BC081628.1 | - | 7 - 8 | 1154 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |