Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mxe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182867.1 | NP_878287.1 | 5 - 6 | 846 | 118 | |
XM_005167664.4 | XP_005167721.2 | 6 - 7 | 978 | 118 | ||
GenBank mRNA | AJ544827.1 | - | 5 - 6 | 846 | 118 | |
BC095587.1 | - | 6 - 7 | 888 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |