Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wnt2ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182876.2 | NP_878296.1 | 2 - 3 | 565 | 80 | |
XM_009302663.3 | XP_009300938.1 | 2 - 3 | 825 | 80 | ||
GenBank mRNA | AF544026.1 | - | 2 - 3 | 568 | 80 | |
BC162076.1 | - | 2 - 3 | 418 | 80 | ||
BC162077.1 | - | 2 - 3 | 418 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |