Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhrs9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199609.1 | NP_955903.1 | 4 - 5 | 826 | 63 | |
GenBank mRNA | BC059614.1 | - | 4 - 5 | 826 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |