Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppdpfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200079.1 | NP_956373.1 | 2 - 3 | 245 | 89 | |
GenBank mRNA | BC057474.1 | - | 2 - 3 | 245 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |