Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: txnipa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200087.1 | NP_956381.1 | 2 - 3 | 548 | 117 | |
GenBank mRNA | BC049493.1 | - | 2 - 3 | 548 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |