Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: laptm4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200098.2 | NP_956392.1 | 4 - 5 | 600 | 139 | |
GenBank mRNA | BC059484.1 | - | 4 - 5 | 529 | 139 | |
EH440716.1 | - | 4 - 5 | 573 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |