Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp2k6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200509.2 | NP_956803.2 | 4 - 5 | 742 | 94 | |
GenBank mRNA | AF283813.1 | - | 4 - 5 | 707 | 94 | |
BC047194.1 | - | 4 - 5 | 728 | 94 | ||
BC055556.1 | - | 4 - 5 | 742 | 94 | ||
BC164859.1 | - | 4 - 5 | 714 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |