Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gchfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200762.3 | NP_957056.1 | 2 - 3 | 224 | 94 | |
XM_005158967.4 | XP_005159024.1 | 2 - 3 | 206 | 94 | ||
GenBank mRNA | BC059590.1 | - | 2 - 3 | 189 | 94 | |
EH535515.1 | - | 1 - 2 | 591 | 94 | ||
EH551858.1 | - | 2 - 3 | 263 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |