Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbm18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200793.1 | NP_957087.1 | 4 - 5 | 491 | 124 | |
GenBank mRNA | BC059651.1 | - | 4 - 5 | 491 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |