Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gipc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200930.1 | NP_957224.1 | 5 - 6 | 1055 | 123 | |
GenBank mRNA | AY989814.1 | - | 5 - 6 | 835 | 123 | |
BC045953.1 | - | 5 - 6 | 1055 | 123 | ||
BC064297.1 | - | 5 - 6 | 1025 | 123 | ||
BC155746.1 | - | 5 - 6 | 840 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |