Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eif3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200999.1 | NP_957293.1 | 4 - 5 | 277 | 70 | |
GenBank mRNA | AY648835.1 | - | 4 - 5 | 254 | 70 | |
BC049046.1 | - | 4 - 5 | 277 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |