Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdcd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201177.1 | NP_957471.1 | 3 - 4 | 426 | 110 | |
GenBank mRNA | BC046067.1 | - | 3 - 4 | 426 | 110 | |
BC071345.1 | - | 3 - 4 | 421 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |