Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ivns1abpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201483.1 | NP_958891.1 | 13 - 14 | 1783 | 89 | |
XM_021479057.1 | XP_021334732.1 | 7 - 8 | 1241 | 89 | ||
GenBank mRNA | BC045449.1 | - | 13 - 14 | 1783 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |