Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ivns1abpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201483.1 | NP_958891.1 | 7 - 8 | 953 | 76 | |
GenBank mRNA | BC045449.1 | - | 7 - 8 | 953 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |