Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pvalb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205572.1 | NP_991135.1 | 1 - 2 | 94 | 72 | |
XM_005164288.3 | XP_005164345.1 | 2 - 3 | 211 | 72 | ||
GenBank mRNA | AF467914.1 | - | 1 - 2 | 94 | 72 | |
BC071552.1 | - | 2 - 3 | 134 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |