Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: irx4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205698.1 | NP_991261.1 | 3 - 4 | 669 | 130 | |
XM_021466666.1 | XP_021322341.1 | 3 - 4 | 765 | 130 | ||
GenBank mRNA | AY845128.1 | - | 3 - 4 | 677 | 130 | |
BC053310.1 | - | 4 - 5 | 620 | 130 | ||
BC065966.1 | - | 3 - 4 | 669 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |